
Coumarin

Synonym(s):
1,2-Benzopyrone, 1-Benzopyran-2-one, 2H-Chromen-2-one, Coumarin
Empirical Formula (Hill Notation):
C9H6O2
CAS Number:
Molecular Weight:
146.14
Products
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing