Skip to Content
Merck
cis-Tramadol-13C, D3 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

cis-Tramadol-13C, D3 hydrochloride solution

cis-Tramadol-13C, D3 hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
13CC15D3H22NO2 · HCl
Molecular Weight:
303.85

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique