
Chiralyst P978

Synonym(s):
Diiodo(p-cymene)ruthenium(II) dimer, Di-μ-iodo(p-cymene)iodoruthenium(II)
Empirical Formula (Hill Notation):
C20H28I4Ru2
CAS Number:
Molecular Weight:
978.19
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing