
Calmodulin-dependent Protein Kinase II fragment 290-309

Synonym(s):
CaM kinase II (290-309)
Empirical Formula (Hill Notation):
C103H185N31O24S
CAS Number:
Molecular Weight:
2273.83
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing