
BDE 209 in 2,2,4-Trimethylpentane

Synonym(s):
Decabromodiphenyl ether solution, Decabromodiphenyl oxide
Empirical Formula (Hill Notation):
C12Br10O
CAS Number:
Molecular Weight:
959.17
Products
Formula Weight
Manufacturer
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing