
Aucubin

Synonym(s):
Aucuboside, Rhimantin
Empirical Formula (Hill Notation):
C15H22O9
CAS Number:
Molecular Weight:
346.33
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing