
Aripiprazole Related Compound C

Synonym(s):
1-(2,3-Dichlorophenyl)piperazine hydrochloride, 1-(2,3-dichlorophenyl)piperazine, 2,3-DCPP HCl
Linear Formula:
C10H12Cl2N2 · HCl
CAS Number:
Molecular Weight:
267.58
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing