
all trans-Retinal

Synonym(s):
Vitamin A aldehyde
Empirical Formula (Hill Notation):
C20H28O
CAS Number:
Molecular Weight:
284.44
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing