
Allopurinol impurity C

Synonym(s):
3-(4H-1,2,4-Triazol-4-yl)-1H-pyrazole-4-carboxamide, 5-(4H-1,2,4-Triazol-4-yl)-1H-pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C6H6N6O
CAS Number:
Molecular Weight:
178.15
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing