
Alachlor

Synonym(s):
2-Chloro-2′,6′-diethyl-N-(methoxymethyl)acetanilide
Empirical Formula (Hill Notation):
C14H20ClNO2
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing