6′-Sialyllactose sodium salt
Synonym(s):
α-NeuNAc-(2→6)-β-D-Gal-(1→4)-D-Glc, 6′-N-Acetylneuraminyl-lactose sodium salt, 6′-SL
Linear Formula:
C23H38NO19Na
CAS Number:
Molecular Weight:
655.53
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Tissue
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing