6-(Boc-amino)hexyl bromide
Synonym(s):
tert-Butyl N-(6-bromohexyl)carbamate
Empirical Formula (Hill Notation):
C11H22BrNO2
CAS Number:
Molecular Weight:
280.20
Products
Product Category
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing