
3-O-Acetyl-11-keto-β-boswellic acid

Synonym(s):
11-keto-β-Boswellic acid acetate, 3α-Acetoxy-11-oxo-12-ursen-24-oic acid, AKBA
Empirical Formula (Hill Notation):
C32H48O5
CAS Number:
Molecular Weight:
512.72
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing