
3,5-Pyrazoledicarboxylic acid monohydrate

Empirical Formula (Hill Notation):
C5H4N2O4 · H2O
CAS Number:
Molecular Weight:
174.11
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing