Skip to Content
Merck
3,5-Dimethoxyaniline 98%

3,5-Dimethoxyaniline

3,5-Dimethoxyaniline 98%
Synonym(s):
5-Aminoresorcinol dimethyl ether
Linear Formula:
(CH3O)2C6H3NH2
CAS Number:
Molecular Weight:
153.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing