3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole
Synonym(s):
1,3-Dihydro-3,3-dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, Togni’s Reagent
Empirical Formula (Hill Notation):
C10H10F3IO
CAS Number:
Molecular Weight:
330.09
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing