Skip to Content
Merck
(1S,2S)-(−)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]ethylenediamine 97%

(1S,2S)-(−)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]ethylenediamine

(1S,2S)-(−)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]ethylenediamine 97%
Synonym(s):
(1S,2S)-(−)-N,N′-Dimethyl-1,2-bis[3-(trifluoromethyl)phenyl]-1,2-ethanediamine
Empirical Formula (Hill Notation):
C18H18F6N2
CAS Number:
Molecular Weight:
376.34

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1