Skip to Content
Merck
1,2-Dioleoyl-3-palmitoyl-rac-glycerol ≥99% (TLC)

1,2-Dioleoyl-3-palmitoyl-rac-glycerol

1,2-Dioleoyl-3-palmitoyl-rac-glycerol ≥99% (TLC)
Synonym(s):
1,2-Di(cis-9-octadecenoyl)-3-hexadecanoyl-rac-glycerol
Empirical Formula (Hill Notation):
C55H102O6
CAS Number:
Molecular Weight:
859.39

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing