1,1-Dioxoisothiazolidine
Synonym(s):
1,1-Dioxo-isothiazolidine
Empirical Formula (Hill Notation):
C3H7NO2S
Molecular Weight:
121.16
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing