Skip to Content
Merck
All Photos(1)

Key Documents

1335202

USP

Hyperoside

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Quercetin 3-D-galactoside, 3,3′,4′,5,7-Pentahydroxyflavone 3-D-galactoside, Hyperin, Hyperoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Beilstein:
5784795
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hyperoside

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15+,17+,18-,21+/m1/s1

InChI key

OVSQVDMCBVZWGM-DTGCRPNFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hyperoside is a naturally occurring flavonoid. It has pharmaceutical properties like anti-inflammatory, anti-viral, anti-oxidative and anti-apoptotic.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Protects against peroxide-induced oxidative damage to cells by scavenging reactive oxygen species and enhancing activity of anti-oxidant enzymes, in particular, catalase and glutathione peroxidase.

Other Notes

A flavonol glycoside widely distribute in plants, including mimosa and St. John′s wort.
Sales restrictions may apply.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC determination of eight polyphenols in the leaves of Crataegus pinnatifida Bge. var. major.
Ying X
Journal of Chromatographic Science, 47(3, 201-205 (2009)
Anti-inflammatory activity of hyperoside through the suppression of nuclear factor-?B activation in mouse peritoneal macrophages.
Kim SJ, Um JY, Lee JY
American Journal of Chinese Medicine, 39(1), 171-181 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service