Skip to Content
Merck
All Photos(1)

Key Documents

SAB2109074

Sigma-Aldrich

Anti-ORAI1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CRACM1, FLJ14466, ORAT1, TMEM142A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity (predicted by homology)

rat, rabbit, yeast, bovine, horse, canine, pig, human

concentration

0.5 mg/mL

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ORAI1(57396)

General description

ORAI1 belongs to the Orai family. ORAI1 (CRACM1) is a plasma membrane protein essential for store-operated calcium entry. Defects in ORAI1 are a cause of severe combined immunodeficiency with CRAC channel dysfunction (CRAC-SCID).CRACM1 is a plasma membrane protein essential for store-operated calcium entry (Vig et al., 2006 [PubMed 16645049]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-331 BC075831.1 1-331 332-844 BC013386.1 220-732 845-990 BC075831.1 845-990 991-1490 BG574128.1 52-551 1491-1496 AK027372.1 1235-1240

Immunogen

Synthetic peptide directed towards the middle region of human ORAI1

Sequence

Synthetic peptide located within the following region: IGTLLFLAEVVLLCWVKFLPLKKQPGQPRPTSKPPASGAAANVSTSGITP

Physical form

Supplied at 0.5 mg/ml in phosphate-buffered saline, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service