Skip to Content
Merck
All Photos(2)

Key Documents

RAB0430

Sigma-Aldrich

Human ShhN ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type urine
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 42 pg/mL
standard curve range: 40.96-10000 pg/mL
(Assay Diluent C)

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 40.96-10000 pg/mL
(Assay Diluent B)

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SHH(6469)

General description

The Human Shh-N (Sonic Hedgehog N Terminus) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme- linked immunosorbent assay for the quantitative measurement of human Shh-N in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human ShhN

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Sonic hedgehog (Shh) is a secreted glycoprotein involved in embryonic morphogenic development of several tissues. It has a role in fracture healing and regeneration. In the canonical pathway, the protein associates with its receptor patched (Ptc-1), de-represses smoothened (Smo) and activates the Gli family of transcription factors. Mutations in the Shh pathway have been linked to developmental diseases and cancer. Shh plays a vital role in axon growth and guidance throughout the brain development. Shh stimulates neurite outgrowth by overcoming neurite growth inhibitory environment associated with glial scar.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonic hedgehog stimulates neurite outgrowth in a mechanical stretch model of reactive-astrogliosis.
Berretta A
Scientific Reports, 6, 21896-21896 (2016)
Expression and Role of Sonic Hedgehog in the Process of Fracture Healing with Aging.
Matsumoto K
In Vivo, 30(2), 99-105 (2016)
Xuqi Li et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(16), 4326-4338 (2014-06-21)
Pancreatic cancer is characterized by stromal desmoplasia and perineural invasion (PNI). We sought to explore the contribution of pancreatic stellate cells (PSC) activated by paracrine Sonic Hedgehog (SHH) in pancreatic cancer PNI and progression. In this study, the expression dynamics
Xuanming Shi et al.
Nature communications, 5, 5425-5425 (2014-11-06)
The Sonic hedgehog (Shh) signalling pathway plays important roles during development and in cancer. Here we report a Shh-induced epigenetic switch that cooperates with Gli to control transcription outcomes. Before induction, poised Shh target genes are marked by a bivalent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service