Skip to Content
Merck
All Photos(2)

Key Documents

HPA051428

Sigma-Aldrich

Anti-DBI antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ACBD1, Anti-ACBP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MSQAEFEKAAEEVRHLKTKPSDEEMLFIYGHYKQATVGD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DBI(1622)

General description

Diazepam binding inhibitor (DBI) is a peptide also known as acyl-coenzyme A binding protein. DBI is majorly expressed in the central nervous system. DBI is also expressed in the peripheral tissues, where there is high affinity for benzodiazepine recognition sites. The gene encoding for DBI is localised on human chromosome 2q14.2.

Immunogen

diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-DBI antibody recognises DBI protein

Biochem/physiol Actions

Diazepam binding inhibitor (DBI) is an active peptide and exerts many functions in the brain and peripheral tissues. DBI displaces diazepam from benzodiazepine and thereby regulates signal transduction at type A gamma amino butyric acid (GABAA) receptors. DBI is also involved in the synthesis of steroids and neuroactives. DBI regulates glucose stimulated insulin secretion. The levels of DBI in cerebrospinal fluid is associated with depression associated with or without anxiety.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST87583

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acyl-CoA-binding protein/diazepam-binding inhibitor gene and pseudogenes: A typical housekeeping gene family
Mandrup S, et al.
Journal of Molecular Biology, 228(3), 1011-1022 (1992)
Association of a Met88Val diazepam binding inhibitor (DBI) gene polymorphism and anxiety disorders with panic attacks
Thoeringer CK, et al.
Journal of Psychiatric Research, 41(7), 579-584 (2007)
Diazepam binding inhibitor (DBI): a peptide with multiple biological actions
Costa E and Guidotti A
Life Sciences, 49(5), 325-344 (1991)
P W Gray et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(19), 7547-7551 (1986-10-01)
Diazepam binding inhibitor (DBI) is a protein that displaces ligands bound to the beta-carboline/benzodiazepine recognition site, an allosteric modulatory site of the type A gamma-aminobutyric acid receptor complex. An incomplete rat cDNA clone coding for DBI was isolated. This rat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service