Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9042-75

illustra Tissue Mini (50)

Cytiva 28-9042-75, sufficient for 50 purifications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

usage

sufficient for 50 purifications

packaging

pkg of 50 ea

manufacturer/tradename

Cytiva 28-9042-75

capacity

>35 μg binding capacity

Related Categories

General description

Tissue and cells genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from animal tissues and mammalian cell cultures, in just 90 min for tissues and 45 min for cells, using DNA spin columns.

Human and animal genomic DNA is found in the chromosomes and it is much longer and higher in molecular weight than plasmid DNA. Genomic DNA is the code for the genome, and this is what is needed to do genome sequencing.

For sequencing it is desirable that the genomic DNA extraction technique used allows the collection of intact genomic DNA in the longest fragments possible. A kit format can help standardize the procedures used to prepare tissue and cell genomic DNA samples for analysis.

To minimize the incidence of DNA shearing during extraction, Tissue and cells genomicPrep Mini Spin Kit includes a lysis solution with proteinase K to gently release genomic DNA from tissues or cells, including human genomic DNA.

The kit can be used for DNA isolation from diverse tissue types such as liver, kidney, and mouse tails with yields ranging from 0.5 to 1.5 μg of genomic DNA per mg of tissue. The kit consistently generates purified genomic DNA with optical density A260/A280 readings of 1.8 or higher.

The gDNA produced can then be further analyzed in PCR, for SNP genotyping, and next generation sequencing (NGS) projects.

Features and Benefits

  • Rapid procedure produces high molecular weight gDNA with minimal shearing
  • Capable of purifying up to 1.5 µg/mg of tissue from an input tissue sample range of 5 to 5NA mg, and up to 4NA µg of genomic DNA per 5 x 1NA6 cells depending on cell type.
  • Total DNA extraction time reduced significantly to 9NA min.
  • Produces high-quality genomic DNA with high molecular weight and less degradation.
  • Verified in common downstream applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Description of common problems that can occur during genomic DNA preparation

Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.

Protocols

This page shows genomic DNA purification from sample applied to FTA cards from Cytiva.

This page shows Genomic DNA purification using illustra™ genomicPrep Mini Spin Kits from Cytiva.

Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service