Skip to Content
Merck
All Photos(1)

Key Documents

CRISPR16H

Sigma-Aldrich

Lenti CRISPR HPRT1 gRNA Control Transduction Particles, High Titer (LV02 vector)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

form

liquid

Quality Level

packaging

vial of 8x25 μL (aliquots)

concentration

(5x108 TU/ml (via p24 titering assay))

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

General description

The Lenti CRISPR HPRT1 gRNA Control Transduction Particles (LV02 vector) product is a critical positive control used to monitor transduction efficiency for lentiCRISPRs. The protein encoded by this gene is a transferase which plays a central role in the generation of purine nucleotides. Cells with intact HPRT1 will die upon exposure to 6-TG (Product No. A4882), an antimetabolite used in the treatment of leukemias. Conversely, cells with HPRT1 knocked out will increase over time with 6-TG selection. This targets a validated HPRT1 CRISPR site, which serves as an experimental control for lenti CRISPRs. The lenti CRISPR positive control uses a dual-component system consisting of a U6-driven guide RNA plasmid targeting the human HPRT1 gene and Ef1a-driven Puromycin resistance casette. This control requires Cas9 be delivered by a separate vector.

Lentiviral-based particles permit efficient infection and integration of the construct into differentiated and non-dividing cells, such as neurons and dendritic cells, overcoming low transfection and integration difficulties when using these cell lines. Self-inactivating replication incompetent viral particles are produced in packaging cells (HEK293T) by co-transfection with compatible packaging and envelope plasmids.

Application

Functional Genomics/Screening/Target Validation

Other Notes

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells, respectively.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service