Skip to Content
Merck
All Photos(1)

Key Documents

CLS353502

Corning® Falcon® Cell Culture Insert Companion Plate

wells, 6, Tissue Culture (TC)-treated surface, sterile

Synonym(s):

Cell Culture Insert Companion Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

material

polystyrene plate

sterility

sterile

feature

lid
wells 6

packaging

case of 50 ea (Individually wrapped)

manufacturer/tradename

Corning 353502

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Falcon® 6 Well TC-Treated Polystyrene Cell Culture Insert Companion Plate, with Lid, Sterile, 1/Pack, 50/Case

Application

Cell Culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service