Skip to Content
Merck
All Photos(1)

Key Documents

Y0001761

Nicorandil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(Pyridine-3-carbonylamino)ethyl nitrate, 2-Nicotinamidoethyl nitrate, Ikorel, N-[2-(Nitrooxy)ethyl]-3-pyridinecarboxamide, SG-75, Sigmart, Zynicor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N3O4
CAS Number:
Molecular Weight:
211.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nicorandil

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[O-][N+](=O)OCCNC(=O)c1cccnc1

InChI

1S/C8H9N3O4/c12-8(7-2-1-3-9-6-7)10-4-5-15-11(13)14/h1-3,6H,4-5H2,(H,10,12)

InChI key

LBHIOVVIQHSOQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nicorandil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Nicorandil is a hybrid ATP-sensitive K+ (KATP) channel opener and nicotinamide nitrate NO donor. Nicorandil selectively activates SUR2B- versus SUR2A-containing KATP channels. It enhances endothelial NO synthase expression and protects against ischemic ventricular arrhythmias. By activating potassium channels, and donating nitric oxide to activate the enzyme guanylate cyclase, Nicorandil causes activation of GMP leading to both arterial and venous vasodilatation. Nicorandil is selective for vascular potassium channels, but has no significant action on cardiac contractility and conduction.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Org. Perox. D

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service