Skip to Content
Merck
All Photos(1)

Key Documents

93856

Supelco

3-Octanol

analytical standard

Synonym(s):

Ethyl pentyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH2CH3
CAS Number:
Molecular Weight:
130.23
Beilstein:
1719310
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

~4.5 (vs air)

vapor pressure

~1 mmHg ( 20 °C)

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.426 (lit.)
n20/D 1.426-1.428

bp

174-176 °C (lit.)

density

0.818 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCC(O)CC

InChI

1S/C8H18O/c1-3-5-6-7-8(9)4-2/h8-9H,3-7H2,1-2H3

InChI key

NMRPBPVERJPACX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Octanol may be used as a reference standard in the determination of the analyte in herb plants using gas chromatography coupled with mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F

Flash Point(C)

68 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Gherman et al.
Talanta, 53(1), 253-262 (2008-10-31)
A rapid extraction procedure for fingerprint chromatogram of different types of Mentha piperita L. (Labiatae family) is presented. The extracts were analysed by gas chromatography and gas chromatography-mass spectrometry (GC/MS). The main volatile compounds identified by the gas chromatography-mass spectrometric
Elena Canellas et al.
Journal of mass spectrometry : JMS, 49(11), 1181-1190 (2014-11-15)
Pressure-sensitive adhesives (PSA) are used to manufacture labels that are applied directly on the food. These adhesives could contain not only intentionally added compounds (IAS) to the adhesive formula but also non-intentionally added substances (NIAS), due to the impurities from
Isabelle Bourdet et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(3), 1043-1051 (2015-01-23)
The APP plays a central role in AD, a pathology that first manifests as a memory decline. Understanding the role of APP in normal cognition is fundamental in understanding the progression of AD, and mammalian studies have pointed to a
Motomi Matsuno et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(14), 5557-5565 (2015-04-10)
Long-term memory (LTM) formation requires de novo gene expression in neurons, and subsequent structural and functional modification of synapses. However, the importance of gene expression in glia during this process has not been well studied. In this report, we characterize
Yoshinori Aso et al.
eLife, 3, e04580-e04580 (2014-12-24)
Animals discriminate stimuli, learn their predictive value and use this knowledge to modify their behavior. In Drosophila, the mushroom body (MB) plays a key role in these processes. Sensory stimuli are sparsely represented by ∼2000 Kenyon cells, which converge onto

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service