Skip to Content
Merck
All Photos(2)

Key Documents

41558

Supelco

Formaldehyde O-pentafluorophenylmethyl-oxime

analytical standard

Synonym(s):

Formaldehyde O-2,3,4,5,6-PFBHA-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4F5NO
CAS Number:
Molecular Weight:
225.12
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

Fc1c(F)c(F)c(CON=C)c(F)c1F

InChI

1S/C8H4F5NO/c1-14-15-2-3-4(9)6(11)8(13)7(12)5(3)10/h1-2H2

InChI key

SRTQFRQWTUMMTC-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Mucci et al.
La Medicina del lavoro, 110(6), 446-458 (2019-12-18)
In recent years, under-vacuum sealing (UVS) and containers with formalin encapsulated in the lid have been proposed for the reduction of occupational exposure to airborne formaldehyde (FA) in healthcare environments. We are presenting a study focused on the assessment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service