Skip to Content
Merck
All Photos(1)

Key Documents

32764

Supelco

Aflatoxin B1-13C17 solution

~0.5 μg/mL in acetonitrile, analytical standard

Synonym(s):

Fully 13C-labelled Aflatoxin B1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C17H12O6
CAS Number:
Molecular Weight:
329.15
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

~0.5 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

mass shift

M+17

storage temp.

−20°C

SMILES string

[13CH3]O[13c]1[13cH][13c]2O[13C@H]3O[13CH]=[13CH][13C@H]3[13c]2[13c]4O[13C](=O)[13C]5=[13C]([13CH2][13CH2][13C]5=O)[13c]14

InChI

1S/C17H12O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h4-6,8,17H,2-3H2,1H3/t8-,17+/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1

InChI key

OQIQSTLJSLGHID-DENWKYDXSA-N

General description

Aflatoxins are a group of hypertoxic secondary metabolites produced by fungi of the genus Aspergillus, mostly A. flavus, A. Parasiticus, and A. nomius. Of the 20 aflatoxins identified so far, B1, B2, G1, and G2 are the four naturally occurring aflatoxins. Aflatoxin B1-13C17 is the 13C isotope-labeled analog of mycotoxin, aflatoxin B1.

Application

Aflatoxin B1-13C17 may be used as an internal standard for the determination of aflatoxin B1 in food grade gums, Pu-erh tea samples, animal feedmilk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).

Analysis Note

purity : ≥97.0% (HPLC)

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple mycotoxins analysis in animal feed with LC-MS/MS: Comparison of extract dilution and immunoaffinity clean-up.
Jedziniak P, et al.
Journal of Separation Science, 10(39), 4776-4783 (2019)
Rapid qualitative and quantitative analysis of aflatoxin B1 in Pu-erh tea by liquid chromatography-isotope dilution tandem mass spectrometry coupled with the QuEChERS purification method.
Dong X, et al.
Analytical Methods : Advancing Methods and Applications, 10(39), 4776-4783 (2018)
Interlaboratory Validation of a Stable Isotope Dilution and Liquid Chromatography Tandem Mass Spectrometry Method for the Determination of Aflatoxins in Milk, Milk-Based Infant Formula, and Feed.
Zhang K, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 101(3), 677-685 (2018)
Kai Zhang et al.
Journal of AOAC International, 97(3), 889-895 (2014-07-24)
Stable isotope dilution with LC/MSIMS was used to determine the following 11 mycotoxins in food grade gums: aflatoxins B1, B2, G1, and G2; deoxynivalenol; fumonisins B1, B2, and B3; ochratoxin A; T-2 toxin; and zearalenone. Samples were fortified with 11

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service