Skip to Content
Merck
All Photos(1)

Key Documents

ZLXUVLP01

Milli-Q

Bactericidal UV lamp

For limiting bacterial growth in Milli-Q®, Elix® and RiOs water systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
NA.21

Quality Level

feature

Bactericidal UV lamp with effective germicidal actions, multidirection radiation

packaging

pkg of 1 unit

UV absorption

λ: 254 nm Amax

compatibility

for use with Elix® Advantage
for use with Elix® Essential UV
for use with Milli-Q® Integral
for use with RiOs-DI® UV
for use with RiOs-DI® Clinical

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The bactericidal UV lamp is a built-in device designed to reduce bacterial levels before the water is stored in a polyethylene (PE) reservoir.

Application

This UV lamp inactivates bacteria.

Features and Benefits

  • 254 nm mercury UV lamp
  • Bactericidal technology

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany
RIOS-DI is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service