Skip to Content
Merck
All Photos(1)

Key Documents

YT30142HW

Millipore

Hazardous Waste Filtration System, 142 mm

USEPA EP Toxicity Test & Toxicity Characteristic Leaching Procedure (TCLP).

Synonym(s):

Waste filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24

material

316 stainless steel
PTFE seal
aluminum leg(s)
molded polypropylene handwheel(s)

Quality Level

parameter

6.9 bar max. inlet pressure (100 psi)

H

42.2 cm (16.6 in.)

diam.

18.4 cm

filter diam.

142 mm

filtration area

120 cm2

weight

12.3 kg (27.1 lb)

fitting

1/8 in. female NPT drain/vent connection
1.5 in. inlet sanitary flange for 14 mm (9/16 in.) I.D. hose (with clamps and adapters)
1.5 in. inlet/outlet sanitary flange for 14 mm (9/16 in.) I.D. hose (with clamps and adapters)
1.5 in. outlet sanitary flange for 14 mm (9/16 in.) I.D. hose (with clamps and adapters)
1/8 in. female NPT upstream port (for vent/relief)

shipped in

ambient

General description

If your waste samples contain non-volatile or semi-volatile substances or metals, use this Hazardous Waste Filtration System for saparation, extratction, and filtration. Developed in collaboration with the US Environmental Protection Agency (EPA), the system is designed specifically for the US EPA Extraction Procedure (EP) Toxicity Test and Toxicity Characteristic Leaching Procedure (TCLP). The device separates solid and liquid phases of waste samples for analysis.



Features & Benefits:

•Contains interior holder with PTFE coating to prevent heavy metal contamination

•Allows easy introduction of bulky samples through wide entry port and removable top plate

•Disassembles for easy cleaning



Applications:

US EPA EP Toxicity Test and Toxicity Characteristic Leaching Procedure



YT31042HW Hazardous Waste Filtration System, 142 mm



Replacement Parts:

YY2214257 Hand Knob

YY2004055 1-1/2 in. TC Silicone Gasket

YY2004045 Clamp 1-1/2 in. TC

YY2214265 O-ring (2-251), silicone

YY2214253 O-ring (2-251), PTFE

YT3014257 Underdrain Support, PTFE-coated

YY2214251 Legs with Caps, 3/16 in. wrench

YY2214252 Handwheel Wrench for 142 mm Holder



Accessories:

YY2004057 Gasket, 1-1/2 in., PTFE-coated fluoroelastomer

XX6700035 Tubing for Pressure Use, 1/2 in. I.D.



Membrane Filters:

AP4014250 Glass Fiber filter without Binder, AP40, 142 mm

APFF14250 Glass Fiber Filter without Binder, APFF, 142 mm



YT310RAHW Rotary Agitator

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service