Key Documents
NA38/26
Millipore
NovAseptic Valve, Diaphragm, Silicone Platinum Cured
About This Item
UNSPSC Code:
31411800
material
platinum diaphragm (Cured Silicone)
silicone (rubber)
Quality Level
feature
hydrophobic (Surface Finish, Exterior)
parameter
-1-6 bar pressure range
-50 °C temp. range (220 °C) (Dry Heat (oxygen rich environment) )
130 °C temp. range (max Steam)
150 °C temp. range (max Dry Heat (oxygen poor environment))
weight
0.09 kg (0.20 lb)
General description
Device Material ELASTOSIL R 4001/70MH Silicone RubberThe Diaphragm is manufactured from 100% medical grade silicone rubber, platinum cured.Chemically, for your specific needs, see appropriate guidelines and actual media characteristics.
NovAseptic Valve, Diaphragm, Silicone Platinum Cured
Application
The applied valve body and actuator may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI