Skip to Content
Merck
All Photos(4)

Key Documents

CFSP22305PK

Millipore

Cellulose Absorbent Sample Pads

thickness 0.83 mm, filter L × W 20 cm × 30 cm, pack of 5 sheets

Synonym(s):

Absorbent pad, CFSP 20 cm x 30 cm sheet, Cellulose pad, Sample pad

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NA.47

packaging

pack of 5 sheets
pkg of 5 sheets

Quality Level

manufacturer/tradename

SureWick®

filter L × W

20 cm × 30 cm

thickness

0.83 mm

General description

SureWick® Pad Materials are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. SureWick® cellulose fiber pad materials are used as sample and absorbant pads in lateral flow assays. This sample pack contains 5 sheets of 20cm x 30cm C083 cellulose fiber pad material.

Application

SureWick® Pad Materials are optimised for use as sample, conjugate or absorbent pads in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing and animal health.

Features and Benefits

  • SureWick® cellulose fiber pads are made from non-woven, 100% pure cellulose fiber.
  • These pads do not contain binders or glues, which can interfere with assay performance.
  • Manufactured to ensure excellent consistency, they provide reliable, predictable sample flow.
  • They exhibit consistent thickness, bed volume, and uniform surface quality.
  • Quick delivery and lead time.
  • Backed by excellent technical support.

Legal Information

SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service