Skip to Content
Merck
All Photos(1)

Key Documents

ABS549

Sigma-Aldrich

Anti-MuSK Antibody

serum, from rabbit

Synonym(s):

Muscle, skeletal receptor tyrosine-protein kinase, Muscle-specific tyrosine-protein kinase receptor, MuSK, Muscle-specific kinase receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MUSK(4593)

General description

MuSK, also known as Muscle, skeletal receptor tyrosine-protein kinase, Muscle-specific tyrosine-protein kinase receptor, or Muscle-specific kinase receptor, and encoded by the gene MUSK, a key muscle specific receptor tyrosine kinase that plays a central role in the formation and maintenance of the neuromuscular junction (NMJ). The NMJ is the special synapse between the motor neuron and the skeletal muscle. MuSK activation in the myotubes regulates the formation of NMJs through the regulation of different processes including the specific expression of genes in the subsynaptic nuclei, the reorganization of the actin cytoskeleton and the clustering of the acetylcholine receptors in the postsynaptic membrane. MuSK also plays a role in the CNS by mediating cholinergic responses, synaptic plasticity and memory function. MuSK is a membrane bound protein localized to the postsynaptic side of the cell membrane at the neuromuscular junction in muscle cells.

Immunogen

Epitope: Extracellular domain
Recombinant protein corresponding to the extracellular domain of mouse MuSK.

Application

Anti-MuSK antibody is an antibody against MuSK for use in western blotting, IHC & IP.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: A representative lot detected MuSK in C2C12 cell lysate (Simeone, L., et al. (2010). The Journal of Neuro. 30(19):6620-6634).
Immunohistochemistry Analysis: A representative lot detected MuSK in mouse soleus muscle tissue (Simeone, L., et al. (2010). The Journal of Neuro. 30(19):6620-6634).
Immunoprecipitation Analysis: A representative lot immunoprecipitated MuSK in C2C12 cell lysate (Simeone, L., et al. (2010). The Journal of Neuro. 30(19):6620-6634).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MuSK in 10 µg of HEK293 cell lysate.

Target description

~110 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bisei Ohkawara et al.
JCI insight, 5(7) (2020-04-10)
Congenital myasthenic syndromes (CMS) are caused by mutations in molecules expressed at the neuromuscular junction. We report clinical, structural, ultrastructural, and electrophysiologic features of 4 CMS patients with 6 heteroallelic variants in AGRN, encoding agrin. One was a 7.9-kb deletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service