Skip to Content
Merck
All Photos(1)

Key Documents

539521

Sigma-Aldrich

Anti-PP2A/Bα (14-27) (PR55) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-PP2A/Bα

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... PTPA(5524)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~55 kDa Bα(PR55α) regulatory subunit of PP2A.

Immunogen

Mammalian
a synthetic peptide (FSQVKGAVDDDVAE) corresponding to amino acids of the mammalian PP2A/Bα regulatory subunit, conjugated to KLH

Application

Immunoblotting (1:500, see comments, see application references)

Immunocytochemistry (1:500)

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM Tris-HCl, 3 mM glycine, pH 7.5.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Reactive with PR55α from most mammalian species, including human, rat, and mouse. Exhibits some cross-reactivity to crude skeletal muscle extracts. Antibody is not recommended for immunoblotting of crude skeletal muscle extracts because of high nonspecific labeling. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service