Skip to Content
Merck
All Photos(1)

Key Documents

491002

Sigma-Aldrich

SAHM1

≥93% (HPLC), solid, Notch transcription factor inhibitor, Calbiochem®

Synonym(s):

Notch Transcription Factor Inhibitor, SAHM1, AcβA1a-ERLRRRI*LCR*HHST (*=S5)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C94H162N36O23S · 7C2HF3O2 · 13H2O
Molecular Weight:
3228.95
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Notch Transcription Factor Inhibitor, SAHM1, Notch Transcription Factor Inhibitor, SAHM1, is a cell-permeable inhibitor that prevents assembly in the NOTCH1 trans-activation complex.

Quality Level

Assay

≥93% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, stabilized hydrocarbon-stapled α-helical peptide that targets a critical protein-protein interface and prevents assembly in the NOTCH1 trans-activation complex. Soluble FITC-SAHM1 was found to bind the pre-formed RAMANK-CSL complex (Kd = 0.12 µM) competitively with MAML1, whereas the unmodified FITC-MAML1 (21-36) peptide bound the complex with markedly diminished affinity in an in vitro pull down assay. Treatment of KOPT-K1 cell lines with 20 µM of compound results in the suppression of NOTCH1 target genes, HES4, DTX1, and HES1. A consistent repressive effect on NOTCH1-activated gene expression by this compound was observed across a panel of human T-ALL cell lines, containing diverse mutant NOTCH1 alleles. In HeLa cells, this peptide also inhibited the expression of ICN1 (IC50 = 6.56 µM) dose-dependently in a luciferase reporter-gene assay, as well as that of β-lactamase (IC50<2.5 µM) in a second reporter-gene assay. Furthermore, treatment with this peptide inhibits NOTCH-mediated proliferation in cultured cells at 15 µM, and in a mouse model of T-ALL cells at 30 mg/kg twice daily.

Warning

Toxicity: Standard Handling (A)

Sequence

Acetyl-βAla-ERLRRRI*LCR*HHST

* = Two S5 molecules (a non-natural alkenyl amino acid) stapled together with a double bond

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Moellering, R.E., et al. 2009. Nature462, 182.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prashan De Zoysa et al.
Development (Cambridge, England), 147(17) (2020-08-12)
The role played by the Notch pathway in cardiac progenitor cell biology remains to be elucidated. Delta-like ligand 4 (Dll4), the arterial-specific Notch ligand, is expressed by second heart field (SHF) progenitors at time-points that are crucial in SHF biology.
Cooperative NF-I?B and Notch1 signaling promotes macrophage-mediated MenaINV expression in breast cancer.
Duran, et al.
Breast Cancer Research, 25, 37-37 (2023)
Ved P Sharma et al.
Nature communications, 12(1), 7300-7300 (2021-12-17)
Cancer stem cells (CSCs) play an important role during metastasis, but the dynamic behavior and induction mechanisms of CSCs are not well understood. Here, we employ high-resolution intravital microscopy using a CSC biosensor to directly observe CSCs in live mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service