Skip to Content
Merck
All Photos(1)

Key Documents

400084

Sigma-Aldrich

HIF Prolyl Hydroxylase Inhibitor

Synonym(s):

HIF Prolyl Hydroxylase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16ClN3O
CAS Number:
Molecular Weight:
361.82
UNSPSC Code:
12352200
NACRES:
NA.54

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

tan

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, hydroxyquinoline HIF Prolyl Hydroxylase (PHD) inhibitor that displays an IC50 = 2 µM in a cell-based assay using a reporter produced by fusing HIF-1α oxygen degradable domain (ODD) to luciferase. This compound (5 µM) is shown to significantly stabilize HIF-1α in vitro and upregulate known HIF target genes, Epo and VEGF, in SH-SY5Y human neuroblastoma cell lines. It also exerts a neuroprotective effect (IC50 = 0.25 µM in an oxidative stress model using cortical neurons.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Smirnova, N.A., et al. 2010. Chem. Biol.4, 380.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Klumpen et al.
Biology of the cell, 109(1), 39-64 (2016-08-16)
Heat stress in ectotherms involves direct (e.g. protein damage) and/or indirect effects (temperature-induced hypoxia and ROS formation), which cause activation of the transcription factors (TF) heat shock factor 1 (HSF-1) and/or hypoxia-inducible factor 1 (HIF-1). The present study focused on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service