Skip to Content
Merck
All Photos(1)

Key Documents

1.44145

Millipore

Power Supply

for use with RCS® High Flow Touch

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

plastic hardware

Agency

non-compendial

feature

electronic

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

storage temp.

room temp

General description

Power Supply is an additional accessory for use with RCS® High Flow Touch microbial air sampler. The RCS® High Flow Touch is a portable air sampler for fast and easy monitoring of ambient air and compressed gases in controlled environments. This microbial air sampler is based on impaction by centrifugal force and can also be used with a compressed gas adapter for additional monitoring of compressed gases. The power supply is used for recharging the Lithium-ion battery of the RCS® High Flow Touch Microbial Air Sampler and Docking station

Application

Power supply is an additional accessory with the RCS® High Flow air sampler for recharging the Lithium-ion battery. These air samplers find application in microbial monitoring of ambient air in the pharmaceutical, medical, and food industries.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service