Skip to Content
Merck
All Photos(1)

Key Documents

1.15093

Supelco

Extrelut® NT Packing Material Refill pack

for column fillings EXtrelut® NT 20

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
272-489-0
NACRES:
NC.07

product name

EXtrelut® NT Refill pack, for column fillings EXtrelut® NT 20

Quality Level

technique(s)

supported liquid extraction (SLE): suitable

pH

10 (20 °C, 100 g/L in H2O)

mp

>450 °C

solubility

0.0037 g/L

density

2.36 g/cm3 at 20 °C

bulk density

300 kg/m3

storage temp.

15-25°C

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

General description

Extrelut® NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut NT1 can take up a maximum of one ml, Extrelut NT3 three ml and Extrelut NT20 up to twenty ml of sample.

Analysis Note

purity test (absorbance 280 nm): ≤ 0.040
purity test (absorbance 250 nm): ≤ 0.080
purity test (absorbance 230 nm): ≤ 0.130

Legal Information

Extrelut is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Simplify your liquid-liquid extractions with our emulsion-free supported-liquid extraction (SLE) single step solution.

Simplify your liquid-liquid extractions with our emulsion-free supported-liquid extraction (SLE) single step solution.

Simplify your liquid-liquid extractions with our emulsion-free supported-liquid extraction (SLE) single step solution.

Simplify your liquid-liquid extractions with our emulsion-free supported-liquid extraction (SLE) single step solution.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service