Key Documents
Z630004
IKA® A 11.2 basic grinding mill cutting blade
A 11.2 cutting blade, 1/cs
About This Item
UNSPSC Code:
41101708
NACRES:
NB.74
material
stainless steel
packaging
pack of 1
case of 1 ×
manufacturer/tradename
IKA 2905200
suitability
suitable for (pulverizing soft, fibrous grinding materials.)
General description
Cutting blade for use with the IKA A11 Basic Analytical Mill, for cutting soft, fibrous ground material. Made of stainless steel
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)