Key Documents
Z266213
Sigma-Aldrich
Polymer-body combination pH electrodes
BNC fitting
About This Item
UNSPSC Code:
41115603
NACRES:
NB.79
stem L × diam.
110 mm × 12 mm
General description
General-purpose, double junction
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance