Skip to Content
Merck
All Photos(1)

Key Documents

SM6050105

Sigma-Aldrich

SmartPor, alumina membrane

thickness 50 μm, pore diameter 60 nm, pore size 125 nm (interpore distance), size 10 mm × 10 mm, pkg of 5 pieces

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al
UNSPSC Code:
12352103
NACRES:
NA.23

description

Pore geometry: trigonal arrangement

packaging

pkg of 5 pieces

size

10 mm × 10 mm

thickness

50 μm

pore size

125 nm (interpore distance)
60 nm pore diameter

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service