Skip to Content
Merck
All Photos(2)

Key Documents

GF79932123

Indium

foil, not light tested, 50x50mm, thickness 0.005mm, temporary acrylic support, 99.8%

Synonym(s):

Indium, IN000080

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In
CAS Number:
Molecular Weight:
114.82
MDL number:
UNSPSC Code:
12141719
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Assay

99.8%

form

foil

manufacturer/tradename

Goodfellow 799-321-23

resistivity

8.37 μΩ-cm

size × thickness

50 x 50 mm × 0.005 mm

mp

156.6 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[In]

InChI

1S/In

InChI key

APFVFJFRJDLVQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loïc Bertrand et al.
The Analyst, 138(16), 4463-4469 (2013-04-05)
In order to fully characterize the zinc white artists' pigment (ZnO), much used since the mid-nineteenth century, three samples collected in the early 20th century were studied using a combination of synchrotron and macroscopic photoluminescence spectroscopy and imaging. An improved
C Takada et al.
Radiation protection dosimetry, 144(1-4), 575-579 (2010-11-11)
At the Nuclear Fuel Cycle Engineering Laboratories of the Japan Atomic Energy Agency (JAEA), small pieces of indium foil incorporated into personal dosemeters have been used for personnel screening in criticality accidents. Irradiation tests of the badges were performed using
Yuri Alexandre Aoto et al.
The Journal of chemical physics, 134(10), 104303-104303 (2011-03-17)
The doubly positively charged gas-phase molecules BrO(2+) and NBr(2+) have been produced by prolonged high-current energetic oxygen (17 keV (16)O(-)) ion surface bombardment (ion beam sputtering) of rubidium bromide (RbBr) and of ammonium bromide (NH(4)Br) powdered ionic salt samples, respectively
[Scintigraphy with indium 111-labeled blood platelets].
C Kessler et al.
Deutsche medizinische Wochenschrift (1946), 109(48), 1853-1859 (1984-11-30)
The significance of chromosomal aberrations in indium-111-labeled lymphocytes.
M L Thakur et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 25(8), 922-927 (1984-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service