Key Documents
BAF133740000
SP Bel-Art® Belpen Marker Pen
black
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
black
manufacturer/tradename
Bel-Art F13374-0000
General description
Felt tip marking pens are perfect for labeling all types of glassware and plastics.
Features and Benefits
  • Permanent, quick drying ink remains waterproof and smudge proof
  • Will withstand autoclaving when dry
  • Pen with handy pocket clip is 13.3cm (5¹/4") long
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP