Skip to Content
Merck
All Photos(1)

Key Documents

933317

Sigma-Aldrich

SEM Sample Holder

Multi Pin Stub Adapter, Jeol

Synonym(s):

Multi Pin Stub Adapte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

EMS techware

Application

This multi pin stub adapter for JEOL 32x11 mm is designed to save time. It is made from vacuum aluminum with stainless steel set screws. It accommodates up to fourt standard 12.7 mm (½") pin stubs with 3.2 mm (⅛") pin.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service