Skip to Content
Merck
All Photos(6)

Documents

921386

Sigma-Aldrich

Reduced graphene oxide enhanced NTCDA composite for battery

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxHyOzNw
UNSPSC Code:
12352119
NACRES:
NA.23

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

Quality Level

sustainability

Greener Alternative Product

application(s)

battery manufacturing

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives, and has been enhanced for energy efficiency. Click here for more information.

Application

Polyimide (PI) is a novel organic material for new generation energy storage devices. PI can be a potential high-power electrode material for rechargeable battery and super-capacitor applications because its faster redox kinetics than inorganic materials. PI-graphene nanocomposite takes advantages of graphene′s excellent electronic conductivity and high surface area, which addresses the issues of PI′s electronic insulation and further improve its utilization. With the added benefit of high safety, flexibility and easy processability, PI composited graphene nanocomposites would penetrate the field of energy storage in the near future.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic Electrode Materials for Rechargeable Lithium Batteries.
Liang Y,et al.
Advanced Energy Materials, 2, 742-769 (2012)
Polyimides: promising energy-storage materials.
Zhiping Song et al.
Angewandte Chemie (International ed. in English), 49(45), 8444-8448 (2010-09-24)
Zhiping Song et al.
Nano letters, 12(5), 2205-2211 (2012-03-28)
Electroactive polymers are a new generation of "green" cathode materials for rechargeable lithium batteries. We have developed nanocomposites combining graphene with two promising polymer cathode materials, poly(anthraquinonyl sulfide) and polyimide, to improve their high-rate performance. The polymer-graphene nanocomposites were synthesized

Articles

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Advanced technologies for energy conversion and storage aim to improve performance and reduce environmental impact.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service