Skip to Content
Merck
All Photos(1)

Key Documents

281239

Sigma-Aldrich

Diallyl carbonate

99%

Synonym(s):

bis(prop-2-enyl) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCH2O)2CO
CAS Number:
Molecular Weight:
142.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

95-97 °C/60 mmHg (lit.)

density

0.991 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCOC(=O)OCC=C

InChI

1S/C7H10O3/c1-3-5-9-7(8)10-6-4-2/h3-4H,1-2,5-6H2

InChI key

JKJWYKGYGWOAHT-UHFFFAOYSA-N

General description

Diallyl carbonate is an acrylating agent. Allyl carbonates are widely employed in Tsuji-Trost allylation, generating carbanion, boronate, phosphide, amide and alkoxide, nucleophiles in situ and improving reaction rates over allyl acetate. Allyl carbonates are also of considerable interest for the intramolecular decarboxylative assymetric couplings.

Application

Transesterification of diallyl carbonate with glycerol in the presence of a catalyst (Pd/PPh3), generates glycerol carbonate.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service