Skip to Content
Merck
All Photos(1)

Key Documents

179590

Sigma-Aldrich

exo-Norborneol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

176-177 °C (lit.)

mp

124-126 °C (lit.)

SMILES string

O[C@H]1C[C@@H]2CC[C@H]1C2

InChI

1S/C7H12O/c8-7-4-5-1-2-6(7)3-5/h5-8H,1-4H2/t5-,6+,7+/m1/s1

InChI key

ZQTYQMYDIHMKQB-VQVTYTSYSA-N

Application

exo-Norborneol was used in the synthesis of cyclic ketones.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective oxidation of alcohols by molecular oxygen over a Pd/MgO catalyst in the absence of any additives.
Green Chemistry, 6(3), 161-165 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service