Skip to Content
Merck
All Photos(1)

Key Documents

15449

Sigma-Aldrich

Boc-His(Trt)-OH

≥98.0% (TLC)

Synonym(s):

Nα-Boc-N(im)-trityl-L-histidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H31N3O4
CAS Number:
Molecular Weight:
497.58
Beilstein:
732035
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

form

powder

optical activity

[α]20/D +12.5±1.0°, c = 1% in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

~130 °C (dec.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](Cc1cn(cn1)C(c2ccccc2)(c3ccccc3)c4ccccc4)C(O)=O

InChI

1S/C30H31N3O4/c1-29(2,3)37-28(36)32-26(27(34)35)19-25-20-33(21-31-25)30(22-13-7-4-8-14-22,23-15-9-5-10-16-23)24-17-11-6-12-18-24/h4-18,20-21,26H,19H2,1-3H3,(H,32,36)(H,34,35)/t26-/m0/s1

InChI key

OYXZPXVCRAAKCM-SANMLTNESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Panayiotis Bilalis et al.
Biomacromolecules, 19(9), 3840-3852 (2018-08-11)
A novel, multifunctional hydrogel that exhibits a unique set of properties for the effective treatment of pancreatic cancer (PC) is presented. The material is composed of a pentablock terpolypeptide of the type PLys- b-(PHIS- co-PBLG)-PLys- b-(PHIS- co-PBLG)- b-PLys, which is
Ngoc-Duc Doan et al.
Journal of peptide science : an official publication of the European Peptide Society, 21(5), 387-391 (2014-11-18)
The solid-phase synthesis of azapeptides possessing a C-terminal aza-residue has been accomplished by a protocol featuring regioselective alkylation of benzhydrylidene-aza-glycinamide and illustrated by the syntheses of [aza-Lys(6)] growth-hormone-releasing peptide-6 analogs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service